Assay Details
| Gene Symbol: APOBEC3Z1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163936.1 | NP_001157408.1 | 2 - 3 | 356 | 81 | |
| GenBank mRNA | EU864534.1 | - | 2 - 3 | 356 | 81 | |
| LC131299.1 | - | 1 - 2 | 139 | 81 | ||
More Information
Set Membership: |
|

APOBEC3Z1
Genomic Map