Assay Details
| Gene Symbol: VIRMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205426.1 | NP_001192355.1 | 17 - 18 | 4248 | 110 | |
| GenBank mRNA | BC133316.1 | - | 3 - 4 | 546 | 110 | |
More Information
Set Membership: |
|

VIRMA
Genomic Map