Assay Details
| Gene Symbol: VIRMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205426.1 | NP_001192355.1 | 23 - 24 | 5304 | 74 | |
| GenBank mRNA | BC133316.1 | - | 9 - 10 | 1602 | 74 | |
More Information
Set Membership: |
|

VIRMA
Genomic Map