Assay Details
| Gene Symbol: PPP1R3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205440.2 | NP_001192369.1 | 3 - 4 | 967 | 122 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPP1R3A| Gene Symbol: PPP1R3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205440.2 | NP_001192369.1 | 3 - 4 | 967 | 122 | |
Set Membership: |
|