Assay Details
| Gene Symbol: RMC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205470.1 | NP_001192399.1 | 11 - 12 | 1146 | 79 | |
| GenBank mRNA | BC151667.1 | - | 11 - 12 | 1118 | 79 | |
More Information
Set Membership: |
|

RMC1
Genomic Map