Assay Details
| Gene Symbol: TMEM62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205497.1 | NP_001192426.1 | 10 - 11 | 1297 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM62| Gene Symbol: TMEM62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205497.1 | NP_001192426.1 | 10 - 11 | 1297 | 78 | |
Set Membership: |
|