Assay Details
| Gene Symbol: CASP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205504.2 | NP_001192433.2 | 3 - 4 | 441 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CASP9| Gene Symbol: CASP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205504.2 | NP_001192433.2 | 3 - 4 | 441 | 57 | |
Set Membership: |
|