Assay Details
Gene Symbol: ZKSCAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205510.3 | NP_001192439.1 | 4 - 5 | 876 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
