Assay Details
| Gene Symbol: DKK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205544.1 | NP_001192473.1 | 1 - 2 | 363 | 83 | |
| GenBank mRNA | JN966753.1 | - | 1 - 2 | 421 | 83 | |
More Information
Set Membership: |
|

DKK1
Genomic Map