Assay Details
| Gene Symbol: UROD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205558.1 | NP_001192487.1 | 5 - 6 | 522 | 65 | |
| GenBank mRNA | BC103285.1 | - | 3 - 4 | 838 | 65 | |
More Information
Set Membership: |
|

UROD
Genomic Map