Assay Details
Gene Symbol: TRIM68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205571.1 | NP_001192500.1 | 3 - 4 | 650 | 64 | |
GenBank mRNA | BC114155.1 | - | 3 - 4 | 649 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
