Assay Details
Gene Symbol: TRIM68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205571.1 | NP_001192500.1 | 6 - 7 | 1029 | 101 | |
GenBank mRNA | BC114155.1 | - | 6 - 7 | 1028 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
