Assay Details
Gene Symbol: BICDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205599.1 | NP_001192528.1 | 4 - 5 | 675 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
