Assay Details
| Gene Symbol: EPOR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205601.1 | NP_001192530.1 | 6 - 7 | 830 | 56 | |
| GenBank mRNA | U61399.1 | - | 5 - 6 | 656 | 56 | |
More Information
Set Membership: |
|

EPOR
Genomic Map