Assay Details
Gene Symbol: SEC62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205604.1 | NP_001192533.1 | 1 - 2 | 315 | 68 | |
GenBank mRNA | BC108162.1 | - | 1 - 2 | 66 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
