Assay Details
Gene Symbol: ZBTB49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205607.1 | NP_001192536.1 | 2 - 3 | 1256 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
