Assay Details
| Gene Symbol: RETREG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205608.2 | NP_001192537.1 | 6 - 7 | 843 | 77 | |
| GenBank mRNA | BT026241.1 | - | 1 - 2 | 149 | 77 | |
More Information
Set Membership: |
|

RETREG2
Genomic Map