Assay Details
Gene Symbol: ETV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205609.1 | NP_001192538.1 | 1 - 2 | 48 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
