Assay Details
| Gene Symbol: MSS51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205641.1 | NP_001192570.1 | 2 - 3 | 812 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MSS51| Gene Symbol: MSS51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205641.1 | NP_001192570.1 | 2 - 3 | 812 | 74 | |
Set Membership: |
|