Assay Details
| Gene Symbol: ZKSCAN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205670.1 | NP_001192599.1 | 1 - 2 | 406 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZKSCAN2| Gene Symbol: ZKSCAN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205670.1 | NP_001192599.1 | 1 - 2 | 406 | 63 | |
Set Membership: |
|