Assay Details
Gene Symbol: ZNF800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205671.1 | NP_001192600.1 | 1 - 2 | 170 | 157 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001205671.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
