Assay Details
| Gene Symbol: ZNF800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205671.1 | NP_001192600.1 | 2 - 3 | 282 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001205671.1 transcripts.
Set Membership: |
|

ZNF800
Genomic Map