Assay Details
| Gene Symbol: AMMECR1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205735.2 | NP_001192664.1 | 2 - 3 | 680 | 60 | |
| GenBank mRNA | BI774580.1 | - | 1 - 2 | 222 | 60 | |
More Information
Set Membership: |
|

AMMECR1L
Genomic Map