Assay Details
Gene Symbol: EBPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205739.1 | NP_001192668.1 | 1 - 2 | 208 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
