Assay Details
| Gene Symbol: ELP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205740.1 | NP_001192669.1 | 6 - 7 | 930 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ELP6| Gene Symbol: ELP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205740.1 | NP_001192669.1 | 6 - 7 | 930 | 59 | |
Set Membership: |
|