Assay Details
| Gene Symbol: ARHGAP45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205741.2 | NP_001192670.2 | 20 - 21 | 2941 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ARHGAP45| Gene Symbol: ARHGAP45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205741.2 | NP_001192670.2 | 20 - 21 | 2941 | 58 | |
Set Membership: |
|