Assay Details
Gene Symbol: FAM21A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205767.2 | NP_001192696.1 | 15 - 16 | 1515 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
