Assay Details
| Gene Symbol: FAM21A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205767.2 | NP_001192696.1 | 16 - 17 | 1639 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM21A| Gene Symbol: FAM21A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205767.2 | NP_001192696.1 | 16 - 17 | 1639 | 97 | |
Set Membership: |
|