Assay Details
| Gene Symbol: IRF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205793.2 | NP_001192722.1 | 8 - 9 | 919 | 60 | |
| GenBank mRNA | AJ490936.1 | - | 6 - 7 | 616 | 60 | |
More Information
Set Membership: |
|

IRF2
Genomic Map