Assay Details
| Gene Symbol: MARCHF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205812.3 | NP_001192741.1 | 23 - 24 | 2380 | 107 | |
| GenBank mRNA | BC119893.1 | - | 13 - 14 | 1453 | 107 | |
| EV632242.1 | - | 2 - 3 | 149 | 107 | ||
More Information
Set Membership: |
|

MARCHF6
Genomic Map