Assay Details
| Gene Symbol: MARCHF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205812.3 | NP_001192741.1 | 25 - 26 | 2653 | 84 | |
| GenBank mRNA | BC119893.1 | - | 15 - 16 | 1726 | 84 | |
| EV632242.1 | - | 4 - 5 | 422 | 84 | ||
More Information
Set Membership: |
|

MARCHF6
Genomic Map