Assay Details
Gene Symbol: EPHA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205822.2 | NP_001192751.2 | 6 - 7 | 1493 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
