Assay Details
| Gene Symbol: SLC17A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205974.1 | NP_001192903.1 | 3 - 4 | 632 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC17A5| Gene Symbol: SLC17A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205974.1 | NP_001192903.1 | 3 - 4 | 632 | 60 | |
Set Membership: |
|