Assay Details
Gene Symbol: EGLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206046.2 | NP_001192975.2 | 4 - 5 | 1322 | 120 | |
GenBank mRNA | DV934069.1 | - | 1 - 2 | 851 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
