Assay Details
Gene Symbol: KAT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206055.1 | NP_001192984.1 | 5 - 6 | 1555 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KAT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206055.1 | NP_001192984.1 | 5 - 6 | 1555 | 74 |
Set Membership: |
![]() ![]() ![]() ![]() |