Assay Details
Gene Symbol: PPIL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206087.2 | NP_001193016.1 | 11 - 12 | 1099 | 69 | |
GenBank mRNA | BC149155.1 | - | 3 - 4 | 237 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
