Assay Details
| Gene Symbol: MAP1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206119.3 | NP_001193048.2 | 1 - 2 | 257 | 56 | |
| GenBank mRNA | BC118407.1 | - | 1 - 2 | 236 | 56 | |
More Information
Set Membership: |
|

MAP1B
Genomic Map