Assay Details
| Gene Symbol: KIF27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206123.3 | NP_001193052.2 | 16 - 17 | 3702 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIF27| Gene Symbol: KIF27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206123.3 | NP_001193052.2 | 16 - 17 | 3702 | 61 | |
Set Membership: |
|