Assay Details
| Gene Symbol: POLR3G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206143.1 | NP_001193072.1 | 4 - 5 | 361 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

POLR3G| Gene Symbol: POLR3G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206143.1 | NP_001193072.1 | 4 - 5 | 361 | 76 | |
Set Membership: |
|