Assay Details
Gene Symbol: NTAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206144.1 | NP_001193073.1 | 8 - 9 | 639 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
