Assay Details
| Gene Symbol: MAP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206151.1 | NP_001193080.1 | 7 - 8 | 1114 | 90 | |
| GenBank mRNA | BC134709.1 | - | 8 - 9 | 1301 | 90 | |
More Information
Set Membership: |
|

MAP9
Genomic Map