Assay Details
| Gene Symbol: RPL34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206221.1 | NP_001193150.1 | 2 - 3 | 185 | 102 | |
| GenBank mRNA | BC103314.1 | - | 2 - 3 | 156 | 102 | |
More Information
Set Membership: |
|

RPL34
Genomic Map