Assay Details
Gene Symbol: CBLIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206239.1 | NP_001193168.1 | 6 - 7 | 908 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CBLIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206239.1 | NP_001193168.1 | 6 - 7 | 908 | 90 |
Set Membership: |
![]() ![]() ![]() |