Assay Details
| Gene Symbol: BATF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192561.2 | NP_001179490.1 | 1 - 2 | 40 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BATF2| Gene Symbol: BATF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192561.2 | NP_001179490.1 | 1 - 2 | 40 | 73 | |
Set Membership: |
|