Assay Details
Gene Symbol: TBX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192630.1 | NP_001179559.1 | 5 - 6 | 841 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
