Assay Details
| Gene Symbol: RORB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192658.1 | NP_001179587.1 | 5 - 6 | 1125 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RORB| Gene Symbol: RORB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192658.1 | NP_001179587.1 | 5 - 6 | 1125 | 65 | |
Set Membership: |
|