Assay Details
| Gene Symbol: GARRE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206284.2 | NP_001193213.2 | 8 - 9 | 1487 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GARRE1| Gene Symbol: GARRE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206284.2 | NP_001193213.2 | 8 - 9 | 1487 | 93 | |
Set Membership: |
|