Assay Details
Gene Symbol: GPR12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206330.2 | NP_001193259.1 | 1 - 2 | 225 | 101 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001206330.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
