Assay Details
| Gene Symbol: SLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206365.1 | NP_001193294.1 | 1 - 2 | 87 | 76 | |
| GenBank mRNA | BC122852.1 | - | 1 - 2 | 81 | 76 | |
| GH327819.1 | - | 1 - 2 | 87 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001206365.1 transcripts.
Set Membership: |
|

SLN
Genomic Map