Assay Details
Gene Symbol: CNNM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206394.2 | NP_001193323.1 | 7 - 8 | 2332 | 56 | |
GenBank mRNA | DV881606.1 | - | 5 - 6 | 614 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
